6-nitro-3,4-dihydro-2H-1,4-benzoxazine ≥95%

Supplier: Matrix Scientific

044382-1G 044382-5G
101865-552EA 172.31 USD
101865-552 101865-554
6-nitro-3,4-dihydro-2H-1,4-benzoxazine ≥95%
6-nitro-3,4-dihydro-2H-1,4-benzoxazine

Hazard: Irritant

Formula: C₈H₈N₂O₃
Melting Pt: 117…119 °C
MDL Number: MFCD07698591
CAS Number: 28226-22-4

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR