Ethyl-2-(4-nitrophenoxy)acetate ≥95%

Supplier: Matrix Scientific

044381-500MG 044381-1G 044381-5G
101865-544EA 181.54 USD
101865-544 101865-546 101865-548
Ethyl-2-(4-nitrophenoxy)acetate ≥95%
Ethyl-2-(4-nitrophenoxy)acetate

Hazard: Irritant

Formula: C₁₀H₁₁NO₅
Melting Pt: 78…80 °C
MDL Number: MFCD00024673
CAS Number: 19076-89-2

Order Now

Specification Test Results

Melting point 78-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR