Methyl-2-phenylimidazo[1,2-a]pyridine-6-carboxylate ≥95%

Supplier: Matrix Scientific

044380-500MG 044380-1G
101865-540EA 538.46 USD
101865-540 101865-542
Methyl-2-phenylimidazo[1,2-a]pyridine-6-carboxylate ≥95%
Methyl 2-phenylimidazo[1,2-a]pyridine-6-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₂N₂O₂
Melting Pt: 181-183°C
MDL Number: MFCD07698590
CAS Number: 962-24-3

Order Now

Specification Test Results

Melting point 181-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR