5-(3-Thienyl)-2-thiophenecarbaldehyde ≥95%

Supplier: Matrix Scientific

044379-500MG 044379-1G
101865-536EA 556.92 USD
101865-536 101865-538
5-(3-Thienyl)-2-thiophenecarbaldehyde ≥95%
5-(3-Thienyl)-2-thiophenecarbaldehyde

Hazard: Irritant

Formula: C₉H₆OS₂
Melting Pt: 96…98 °C
MDL Number: MFCD06740285
CAS Number: 60141-28-8

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR