α-Oxothiophen-2-acetic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Oxo-2-(thiophen-2-yl)acetic acid
044371-500MG
044371-1G
044371-5G
101865-506EA
61.54
USD
101865-506
101865-508
101865-510
α-Oxothiophen-2-acetic acid ≥95%
α-Oxothiophen-2-acetic acid
Hazard: Irritant
Formula:
C₆H₄O₃S MW: 156.1600 g/mol Melting Pt: 96…100 °C |
MDL Number:
MFCD00005441 CAS Number: 4075-59-6 UN: 3261 ADR: 8,III |
Specification Test Results
Melting point | 96-100°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...