3-(4-Chlorophenyl)-2-cyanoacrylic acid ≥95%
Supplier: Matrix Scientific
044363-500MG
044363-1G
044363-5G
101865-482EA
181.54
USD
101865-482
101865-484
101865-486
3-(4-Chlorophenyl)-2-cyanoacrylic acid ≥95%
3-(4-Chlorophenyl)-2-cyanoacrylic acid
Hazard: Irritant
Formula:
C₁₀H₆ClNO₂ MW: 207.62 g/mol Melting Pt: 191…193 °C |
MDL Number:
MFCD02317172 CAS Number: 20374-46-3 |
Specification Test Results
Melting point | 191-193°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...