3-(4-Chlorophenyl)-1H-pyrazol-5-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Amino-5-(4-chlorophenyl)pyrazole, 5-Amino-3-(4-chlorophenyl)-1H-pyrazole, 5-(4-Chlorophenyl)-1H-pyrazol-3-amine
044361-500MG
044361-1G
044361-5G
101865-470EA
278.46
USD
101865-470
101865-472
101865-474
3-(4-Chlorophenyl)-1H-pyrazol-5-amine ≥95%
3-(4-Chlorophenyl)-1H-pyrazol-5-amine
Hazard: Irritant
Formula:
C₉H₈ClN₃ MW: 193.64 g/mol Melting Pt: 171-172°C |
MDL Number:
MFCD00053046 CAS Number: 78583-81-0 |
Specification Test Results
Melting point | 171-172°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...