Methyl-4-bromo-1-(phenylsulfonyl)-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

044359-500MG 044359-1G 044359-5G
101865-458EA 556.92 USD
101865-458 101865-460 101865-462
Methyl-4-bromo-1-(phenylsulfonyl)-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-bromo-1-(phenylsulfonyl)-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀BrNO₄S
Melting Pt: 137…139 °C
MDL Number: MFCD08064739
CAS Number: 882747-46-8

Order Now

Specification Test Results

Melting point 137-139°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR