5-Nitro-2-phenylpyridine ≥95%
Supplier: Matrix Scientific
044353-500MG
044353-1G
044353-5G
101865-424EA
278.46
USD
101865-424
101865-426
101865-428
5-Nitro-2-phenylpyridine ≥95%
5-Nitro-2-phenylpyridine
Hazard: Irritant
Formula:
C₁₁H₈N₂O₂ Melting Pt: 116…118 °C |
MDL Number:
MFCD04114193 CAS Number: 89076-64-2 |
Specification Test Results
Melting point | 116-118°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...