2-(4-Formylphenyl)isonicotinonitrile ≥95%
Supplier: Matrix Scientific
044352-500MG
044352-1G
044352-5G
101865-418EA
556.92
USD
101865-418
101865-420
101865-422
2-(4-Formylphenyl)isonicotinonitrile ≥95%
2-(4-Formylphenyl)isonicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Hazard: Irritant
Formula:
C₁₃H₈N₂O Melting Pt: 175…178 °C |
MDL Number:
MFCD06200959 CAS Number: 253801-11-5 |
Specification Test Results
Melting point | 175-178°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...