2-(4-Methoxyphenyl)isonicotinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-Methoxyphenyl)pyridine-4-carbonitrile

044351-500MG 044351-1G 044351-5G
101865-412EA 747.69 USD
101865-412 101865-414 101865-416
2-(4-Methoxyphenyl)isonicotinonitrile ≥95%
2-(4-Methoxyphenyl)isonicotinonitrile

Hazard: Irritant

Formula: C₁₃H₁₀N₂O
Melting Pt: 97…99 °C
MDL Number: MFCD07366546
CAS Number: 885949-66-6

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR