2-Phenylisonicotinonitrile ≥95%
Supplier: Matrix Scientific
044350-500MG
044350-1G
044350-5G
101865-406EA
556.92
USD
101865-406
101865-408
101865-410
2-Phenylisonicotinonitrile ≥95%
2-Phenylisonicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Hazard: Irritant
Formula:
C₁₂H₈N₂ Melting Pt: 78…80 °C |
MDL Number:
MFCD06200958 CAS Number: 33744-17-1 |
Specification Test Results
Melting point | 78-80°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...