2-(4-Chlorophenyl)-3-nitropyridine ≥95%
Supplier: Matrix Scientific
044349-500MG
044349-1G
044349-5G
101865-400EA
375.38
USD
101865-400
101865-402
101865-404
2-(4-Chlorophenyl)-3-nitropyridine ≥95%
2-(4-Chlorophenyl)-3-nitropyridine
Hazard: Irritant
Formula:
C₁₁H₇ClN₂O₂ Melting Pt: 114…116 °C |
MDL Number:
MFCD06200957 CAS Number: 886361-74-6 |
Specification Test Results
Melting point | 114-116°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...