5-(Chloromethyl)-2-(4-methoxyphenyl)pyridine ≥95%

Supplier: Matrix Scientific

044347-500MG 044347-1G 044347-5G
101865-388EA 556.92 USD
101865-388 101865-390 101865-392
5-(Chloromethyl)-2-(4-methoxyphenyl)pyridine ≥95%
5-(Chloromethyl)-2-(4-methoxyphenyl)pyridine

Hazard: Irritant

Formula: C₁₃H₁₂ClNO
Melting Pt: 46…49 °C
MDL Number: MFCD06200955
CAS Number: 886361-72-4

Order Now

Specification Test Results

Melting point 46-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR