4-[5-(Chloromethyl)-2-pyridinyl]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

044346-500MG 044346-1G 044346-5G
101865-382EA 556.92 USD
101865-382 101865-384 101865-386
4-[5-(Chloromethyl)-2-pyridinyl]benzenecarbaldehyde ≥95%
4-[5-(Chloromethyl)-2-pyridinyl]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₀ClNO
MW: 231.68 g/mol
Melting Pt: 90…92 °C
MDL Number: MFCD06200954
CAS Number: 886361-70-2

Order Now

Specification Test Results

Melting point 90-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR