6-(tert-Butyl)-3-methyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
6-tert-Butyl 3-methyl 2-amino-4,5-dihydrothieno[2,3-c]pyridine-3,6(7H)-dicarboxylate, Methyl 2-amino-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate, N6-BOC protected
044342-500MG
044342-1G
044342-5G
101865-366EA
160
USD
101865-366
101865-368
101865-370
6-(tert-Butyl)-3-methyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate ≥97%
6-(tert-Butyl)-3-methyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
Hazard: Irritant
Formula:
C₁₄H₂₀N₂O₄S MW: 312.39 g/mol Melting Pt: 165…166 °C |
MDL Number:
MFCD06200952 CAS Number: 877041-47-9 |
Specification Test Results
Melting point | 165-166°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...