6-[(tert-Butoxycarbonyl)amino]nicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Aminonicotinic acid, 6-BOC protected, 6-((tert-Butoxycarbonyl)amino)nicotinic acid

044341-1G 044341-250MG
101865-364EA 289.23 USD
101865-364 102551-330
6-[(tert-Butoxycarbonyl)amino]nicotinic acid ≥95%
6-[(tert-Butoxycarbonyl)amino]nicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₄
MW: 238.25 g/mol
Melting Pt: 292…296 °C
MDL Number: MFCD02682388
CAS Number: 231958-14-8

Order Now

Specification Test Results

Melting point 292-296°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR