1,4-Dioxa-9-thia-12-azadispiro[4.2.4.2]tetradecane-11-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
12-Aza-1,4-dioxa-9-thiadispiro[4.2.4.2]tetradecane-11-carboxylic acid
044339-500MG
044339-1G
044339-5G
101865-356EA
181.54
USD
101865-356
101865-358
101865-360
1,4-Dioxa-9-thia-12-azadispiro[4.2.4.2]tetradecane-11-carboxylic acid ≥95%
1,4-Dioxa-9-thia-12-azadispiro[4.2.4.2]tetradecane-11-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₇NO₄S MW: 259.33 g/mol Melting Pt: 201 °C |
MDL Number:
MFCD06411612 CAS Number: 1030253-98-5 |
Specification Test Results
Melting point | 201°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...