4'-Benzyloxy-3'-fluoroacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-(Benzyloxy)-3-fluorophenyl)ethanone

044337-500MG 044337-1G 044337-5G
101865-344EA 556.92 USD
101865-344 101865-346 101865-348
4'-Benzyloxy-3'-fluoroacetophenone ≥95%
4'-Benzyloxy-3'-fluoroacetophenone

Hazard: Irritant

Formula: C₁₅H₁₃FO₂
MW: 244.261 g/mol
Melting Pt: 82-85°C
MDL Number: MFCD06659550
CAS Number: 81227-99-8

Order Now

Specification Test Results

Melting point 82-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR