Methyl-2-(3-amino-4-hydroxyphenyl)acetate ≥95%
Supplier: Matrix Scientific
044333-500MG
044333-1G
044333-5G
101865-332EA
556.92
USD
101865-332
101865-334
101865-336
Methyl-2-(3-amino-4-hydroxyphenyl)acetate ≥95%
Methyl-2-(3-amino-4-hydroxyphenyl)acetate
Hazard: Irritant
Formula:
C₉H₁₁NO₃ Melting Pt: 102…105 °C |
MDL Number:
MFCD04125316 CAS Number: 78587-72-1 |
Specification Test Results
Melting point | 102-105°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...