Ethyl pyrrole-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 1H-pyrrole-2-carboxylate

044331-5G 044331-25G
101865-328EA 20 USD
101865-328 101865-330
Ethyl pyrrole-2-carboxylate ≥97%
Ethyl pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₇H₉NO₂
MW: 139.16 g/mol
Melting Pt: 36…38 °C
MDL Number: MFCD00817049
CAS Number: 2199-43-1

Order Now

Specification Test Results

Melting point 36-38°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR