3-Chloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline ≥95%

Supplier: Matrix Scientific

044327-500MG 044327-1G 044327-5G
101865-310EA 278.46 USD
101865-310 101865-312 101865-314
3-Chloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline ≥95%
3-Chloro-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline

Hazard: Irritant

Formula: C₁₂H₇Cl₂F₃N₂O
Melting Pt: 90…92 °C
MDL Number: MFCD04117811
CAS Number: 76471-06-2

Order Now

Specification Test Results

Melting point 90-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR