Methyl-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

044326-500MG 044326-1G 044326-5G
101865-304EA 375.38 USD
101865-304 101865-306 101865-308
Methyl-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]benzenecarboxylate ≥95%
Methyl-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]benzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₉ClF₃NO₂
Melting Pt: 88…91 °C
MDL Number: MFCD06797388
CAS Number: 885949-65-5

Order Now

Specification Test Results

Melting point 88-91°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR