tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-2-hydroxymethylpiperidine, Tert-Butyl 2-(Hydroxymethyl)Tetrahydro-1(2H)-Pyridinecarboxylate, (±)-1-Boc-2-(hydroxymethyl)piperidine, BOC-2-piperidylmethanol

044324-1G 044324-5G
101865-294EA 106.15 USD
101865-294 101865-296
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate ≥95%
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₁NO₃
Melting Pt: 74…77 °C
MDL Number: MFCD04114966
CAS Number: 157634-00-9

Order Now

Specification Test Results

Melting point 74-77°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR