2-[(4-Chlorophenyl)sulfanyl]-N'-hydroxy-3-pyridinecarboximidamide ≥95%

Supplier: Matrix Scientific

044321-500MG 044321-1G 044321-5G
101865-282EA 556.92 USD
101865-282 101865-284 101865-286
2-[(4-Chlorophenyl)sulfanyl]-N'-hydroxy-3-pyridinecarboximidamide ≥95%
2-[(4-Chlorophenyl)sulfanyl]-N'-hydroxy-3-pyridinecarboximidamide

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃OS
MW: 279.75 g/mol
Melting Pt: 190…192 °C
MDL Number: MFCD05021868
CAS Number: 886360-64-1

Order Now

Specification Test Results

Melting point 190-192°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR