1-Methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carbohydrazide ≥95%
Supplier: Matrix Scientific
044320-500MG
044320-1G
044320-5G
101865-276EA
747.69
USD
101865-276
101865-278
101865-280
1-Methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carbohydrazide ≥95%
1-Methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carbohydrazide
Hazard: Irritant
Formula:
C₈H₇F₃N₄OS Melting Pt: 218…220 °C |
MDL Number:
MFCD04973722 CAS Number: 886360-63-0 |
Specification Test Results
Melting point | 218-220°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...