2-[(4-Chlorophenyl)sulfanyl]nicotinonitrile ≥95%

Supplier: Matrix Scientific

044319-500MG 044319-1G 044319-5G
101865-270EA 375.38 USD
101865-270 101865-272 101865-274
2-[(4-Chlorophenyl)sulfanyl]nicotinonitrile ≥95%
2-[(4-Chlorophenyl)sulfanyl]nicotinonitrile

Hazard: Irritant

Formula: C₁₂H₇ClN₂S
Melting Pt: 112…116 °C
MDL Number: MFCD04113614
CAS Number: 886360-62-9

Order Now

Specification Test Results

Melting point 112-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR