4-[(4-Chlorophenyl)sulfanyl]-3-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(4-Chlorophenylthio)-3-nitrobenzaldehyde

044318-500MG 044318-1G 044318-5G
101865-264EA 90.77 USD
101865-264 101865-266 101865-268
4-[(4-Chlorophenyl)sulfanyl]-3-nitrobenzenecarbaldehyde ≥95%
4-[(4-Chlorophenyl)sulfanyl]-3-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₈ClNO₃S
MW: 293.73 g/mol
Melting Pt: 118…120 °C
MDL Number: MFCD00098773
CAS Number: 270262-96-9

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR