1-Methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

044316-500MG 044316-1G 044316-5G
101865-252EA 1160 USD
101865-252 101865-254 101865-256
1-Methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carbaldehyde ≥95%
1-Methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carbaldehyde

Hazard: Irritant

Formula: C₈H₅F₃N₂OS
Melting Pt: 97…100 °C
MDL Number: MFCD04973721
CAS Number: 886360-61-8

Order Now

Specification Test Results

Melting point 97-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR