2-(4-Nitrophenoxy)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

044315-500MG 044315-1G 044315-5G
101865-246EA 269.23 USD
101865-246 101865-248 101865-250
2-(4-Nitrophenoxy)benzenecarboxylic acid ≥95%
2-(4-Nitrophenoxy)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₉NO₅
Melting Pt: 154…157 °C
MDL Number: MFCD01002507
CAS Number: 6082-87-7

Order Now

Specification Test Results

Melting point 154-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR