Methyl-2-(4-aminophenoxy)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

044314-500MG 044314-1G 044314-5G
101865-240EA 375.38 USD
101865-240 101865-242 101865-244
Methyl-2-(4-aminophenoxy)benzenecarboxylate ≥95%
Methyl-2-(4-aminophenoxy)benzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₃NO₃
Melting Pt: 75…78 °C
MDL Number: MFCD04117809
CAS Number: 212189-58-7

Order Now

Specification Test Results

Melting point 75-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR