Methyl-2-(4-formyl-2-nitrophenoxy)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

044312-500MG 044312-1G 044312-5G
101865-228EA 278.46 USD
101865-228 101865-230 101865-232
Methyl-2-(4-formyl-2-nitrophenoxy)benzenecarboxylate ≥95%
Methyl-2-(4-formyl-2-nitrophenoxy)benzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₁NO₆
Melting Pt: 70…75 °C
MDL Number: MFCD04117808
CAS Number: 886360-60-7

Order Now

Specification Test Results

Melting point 70-75°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR