3-[3-(Trifluoromethyl)phenyl]-1H-pyrazole ≥95%

Supplier: Matrix Scientific

044308-500MG 044308-1G 044308-5G
101865-204EA 169.23 USD
101865-204 101865-206 101865-208
3-[3-(Trifluoromethyl)phenyl]-1H-pyrazole ≥95%
3-[3-(Trifluoromethyl)phenyl]-1H-pyrazole

Hazard: Irritant

Formula: C₁₀H₇F₃N₂
Melting Pt: 78…79 °C
MDL Number: MFCD00973825
CAS Number: 149739-48-0

Order Now

Specification Test Results

Melting point 78-79°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR