3-(Dimethylamino)-1-[3-(trifluoromethyl)phenyl]-2-propen-1-one ≥95%

Supplier: Matrix Scientific

044307-500MG 044307-1G 044307-5G
101865-198EA 181.54 USD
101865-198 101865-200 101865-202
3-(Dimethylamino)-1-[3-(trifluoromethyl)phenyl]-2-propen-1-one ≥95%
3-(Dimethylamino)-1-[3-(trifluoromethyl)phenyl]-2-propen-1-one

Hazard: Irritant

Formula: C₁₂H₁₂F₃NO
Melting Pt: 61…62 °C
MDL Number: MFCD01570011
CAS Number: 72851-19-5

Order Now

Specification Test Results

Melting point 61-62°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR