5-Bromo-4,6-dimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

044303-500MG 044303-1G 044303-5G
101865-182EA 78.46 USD
101865-182 101865-184 101865-186
5-Bromo-4,6-dimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
5-Bromo-4,6-dimethyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₈H₇BrN₂O
MW: 227.07 g/mol
Melting Pt: 263…266 °C
MDL Number: MFCD01117224
CAS Number: 23819-87-6

Order Now

Specification Test Results

Melting point 263-266°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR