3-Methyl-5-(2-thienyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

044302-500MG 044302-1G 044302-5G
101865-176EA 123.08 USD
101865-176 101865-178 101865-180
3-Methyl-5-(2-thienyl)-1H-pyrazole ≥95%
3-Methyl-5-(2-thienyl)-1H-pyrazole

Hazard: Irritant

Formula: C₈H₈N₂S
MW: 164.23 g/mol
Melting Pt: 140…141 °C
MDL Number: MFCD00514532
CAS Number: 31618-80-1

Order Now

Specification Test Results

Melting point 140-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR