5-(5-Chloro-2-thienyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

044301-500MG 044301-1G 044301-5G
101865-170EA 278.46 USD
101865-170 101865-172 101865-174
5-(5-Chloro-2-thienyl)-1H-pyrazole ≥95%
5-(5-Chloro-2-thienyl)-1H-pyrazole

Hazard: Irritant

Formula: C₇H₅ClN₂S
MW: 184.65 g/mol
Melting Pt: 168…169 °C
MDL Number: MFCD03791215
CAS Number: 166196-61-8

Order Now

Specification Test Results

Melting point 168-169°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR