3-(Dimethylamino)-1-(2-pyrazinyl)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Dimethylamino)-1-(pyrazin-2-yl)prop-2-en-1-one

044299-500MG 044299-1G 044299-5G
101865-158EA 556.92 USD
101865-158 101865-160 101865-162
3-(Dimethylamino)-1-(2-pyrazinyl)-2-propen-1-one ≥95%
3-(Dimethylamino)-1-(2-pyrazinyl)-2-propen-1-one

Hazard: Irritant

Formula: C₉H₁₁N₃O
MW: 177.21 g/mol
Melting Pt: 131…133 °C
MDL Number: MFCD02325230
CAS Number: 111781-53-4

Order Now

Specification Test Results

Melting point 131-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR