3-(4-Ethoxy-3-methoxyphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid ≥95%

Supplier: Matrix Scientific

044298-500MG 044298-1G 044298-5G
101865-152EA 278.46 USD
101865-152 101865-154 101865-156
3-(4-Ethoxy-3-methoxyphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid ≥95%
3-(4-Ethoxy-3-methoxyphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid

Hazard: Irritant

Formula: C₂₀H₂₁NO₅
Melting Pt: 190…192 °C
MDL Number: MFCD03791214
CAS Number: 885951-50-8

Order Now

Specification Test Results

Melting point 190-192°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR