4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one ≥95%

Supplier: Matrix Scientific

044294-500MG 044294-1G 044294-5G
101865-140EA 556.92 USD
101865-140 101865-142 101865-144
4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one ≥95%
4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one

Hazard: Irritant

Formula: C₆H₁₁N₃O
MW: 141.17 g/mol
Melting Pt: 230…233 °C
MDL Number: MFCD00053049
CAS Number: 7032-17-9

Order Now

Specification Test Results

Melting point 230-233°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR