Methyl-3-(2-aminophenoxy)-2-thiophenecarboxylate ≥95%
Supplier: Matrix Scientific
044293-500MG
044293-1G
044293-5G
101865-134EA
556.92
USD
101865-134
101865-136
101865-138
Methyl-3-(2-aminophenoxy)-2-thiophenecarboxylate ≥95%
Methyl-3-(2-aminophenoxy)-2-thiophenecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₁NO₃S Melting Pt: 107…109 °C |
MDL Number:
MFCD04117806 CAS Number: 91041-21-3 |
Specification Test Results
Melting point | 107-109°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...