1,4,5,6-Tetrahydro-3-pyridinecarboxamide ≥95%

Supplier: Matrix Scientific

044291-500MG 044291-1G 044291-5G
101865-124EA 230.77 USD
101865-124 101865-126 101865-128
1,4,5,6-Tetrahydro-3-pyridinecarboxamide ≥95%
1,4,5,6-Tetrahydro-3-pyridinecarboxamide

Hazard: Irritant

Formula: C₆H₁₀N₂O
MW: 126.16 g/mol
Melting Pt: 207…210 °C
MDL Number: MFCD00051500
CAS Number: 7032-11-3

Order Now

Specification Test Results

Melting point 207-210°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR