tert-Butyl-4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(4-BOC-Pyrazino)benzylamine, 4-[(Aminomethyl)phenyl]piperazine, N1-BOC protected, tert-Butyl 4-(4-(aminomethyl)phenyl)piperazine-1-carboxylate

044290-500MG 044290-1G 044290-5G
101865-118EA 270.77 USD
101865-118 101865-120 101865-122
tert-Butyl-4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₅N₃O₂
MW: 291.4 g/mol
Melting Pt: 95…96 °C
MDL Number: MFCD03791213
CAS Number: 852180-47-3

Order Now

Specification Test Results

Melting point 95-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR