Methyl-3-(2-amino-4-fluorophenoxy)-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

044287-500MG 044287-1G 044287-5G
101865-112EA 556.92 USD
101865-112 101865-114 101865-116
Methyl-3-(2-amino-4-fluorophenoxy)-2-thiophenecarboxylate ≥95%
Methyl-3-(2-amino-4-fluorophenoxy)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₀FNO₃S
Melting Pt: 106…108 °C
MDL Number: MFCD04117805
CAS Number: 886360-59-4

Order Now

Specification Test Results

Melting point 106-108°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR