tert-Butyl-4-{4-[amino(hydroxyimino)methyl]phenyl}tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

044284-500MG 044284-1G 044284-5G
101865-094EA 556.92 USD
101865-094 101865-096 101865-098
tert-Butyl-4-{4-[amino(hydroxyimino)methyl]phenyl}tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-{4-[amino(hydroxyimino)methyl]phenyl}tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₄N₄O₃
MW: 320.39 g/mol
Melting Pt: 206…207 °C
MDL Number: MFCD05021842
CAS Number: 186650-24-8

Order Now

Specification Test Results

Melting point 206-207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR