N-[(3-Methoxybenzyl)oxy]-1-hydrazinecarboxamide ≥95%

Supplier: Matrix Scientific

044282-500MG 044282-1G 044282-5G
101865-082EA 747.69 USD
101865-082 101865-084 101865-086
N-[(3-Methoxybenzyl)oxy]-1-hydrazinecarboxamide ≥95%
N-[(3-Methoxybenzyl)oxy]-1-hydrazinecarboxamide

Hazard: Irritant

Formula: C₉H₁₃N₃O₃
Melting Pt: 100…103 °C
MDL Number: MFCD03791211
CAS Number: 885951-49-5

Order Now

Specification Test Results

Melting point 100-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR