Methyl-5-chloro-6-(phenylsulfanyl)nicotinate ≥95%

Supplier: Matrix Scientific

044281-500MG 044281-1G 044281-5G
101865-076EA 423.08 USD
101865-076 101865-078 101865-080
Methyl-5-chloro-6-(phenylsulfanyl)nicotinate ≥95%
Methyl-5-chloro-6-(phenylsulfanyl)nicotinate

Hazard: Irritant

Formula: C₁₃H₁₀ClNO₂S
Melting Pt: 100…103 °C
MDL Number: MFCD03791210
CAS Number: 885951-48-4

Order Now

Specification Test Results

Melting point 100-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR