Methyl-4-hydroxy-5-oxo-1-(2-pyridinylmethyl)-2,5-dihydro-1H-pyrrole-3-carboxylate ≥95%

Supplier: Matrix Scientific

044280-500MG 044280-1G 044280-5G
101865-070EA 278.46 USD
101865-070 101865-072 101865-074
Methyl-4-hydroxy-5-oxo-1-(2-pyridinylmethyl)-2,5-dihydro-1H-pyrrole-3-carboxylate ≥95%
Methyl-4-hydroxy-5-oxo-1-(2-pyridinylmethyl)-2,5-dihydro-1H-pyrrole-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₄
MW: 248.24 g/mol
Melting Pt: 148…149 °C
MDL Number: MFCD03791209
CAS Number: 885951-47-3

Order Now

Specification Test Results

Melting point 148-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR