6-Chloro-2H-[1,4]benzoxazine-3(4H)-one ≥95%

Supplier: Matrix Scientific

044279-500MG 044279-1G 044279-5G
101865-064EA 181.54 USD
101865-064 101865-066 101865-068
6-Chloro-2H-[1,4]benzoxazine-3(4H)-one ≥95%
6-Chloro-2H-[1,4]benzoxazine-3(4H)-one

Hazard: Irritant

Formula: C₈H₆ClNO₂
Melting Pt: 217…219 °C
MDL Number: MFCD00174511
CAS Number: 7652-29-1

Order Now

Specification Test Results

Melting point 217-219°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR