1-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-4-ol ≥95%

Supplier: Matrix Scientific

044278-500MG 044278-1G 044278-5G
101865-058EA 375.38 USD
101865-058 101865-060 101865-062
1-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-4-ol ≥95%
1-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-4-ol

Hazard: Irritant

Formula: C₁₀H₇F₃N₂O
Melting Pt: 99…101 °C
MDL Number: MFCD03791208
CAS Number: 77458-38-9

Order Now

Specification Test Results

Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR